Assay Details
| Gene Symbol: NEXN-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103535.1 | 2 | 1657 | 85 | ||
| GenBank mRNA | AK075118.1 | - | 3 | 1638 | 85 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Oligo Microarray 4x44K | 33651 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 29701 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 9019 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P110780 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3395219 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P110780 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7917153 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7917152 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7917154 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7917155 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7917151 | - | - |
| Affymetrix HT HG-U133+ PM Array Plate | 1560746_PM_at | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006748.hg.1 | ENST00000503186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006748.hg.1 | ENST00000503186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006748.hg.1 | ENST00000503186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006748.hg.1 | ENST00000503186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006748.hg.1 | ENST00000503186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006748.hg.1 | ENST00000503186.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006748.hg.1 | ENST00000509453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006748.hg.1 | ENST00000509453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400006748.hg.1 | ENST00000509453.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | ENST00000597757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | ENST00000597757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | ENST00000597757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | ENST00000597757.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100014659.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209350.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209350.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209350.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209350.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209350.hg.1 | TC0100014659.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100209349.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209349.hg.1 | ENST00000597757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209349.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209349.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209349.hg.1 | ENST00000597757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209349.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209349.hg.1 | TC0100014659.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0400006748.hg.1 | ENST00000503186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006748.hg.1 | ENST00000503186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006748.hg.1 | ENST00000503186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006748.hg.1 | ENST00000503186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006748.hg.1 | ENST00000503186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006748.hg.1 | ENST00000503186.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006748.hg.1 | ENST00000509453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006748.hg.1 | ENST00000509453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400006748.hg.1 | ENST00000509453.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209356.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209356.hg.1 | ENST00000597757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209356.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209356.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209356.hg.1 | ENST00000597757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100209356.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100209356.hg.1 | TC0100014659.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | ENST00000421331.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | ENST00000597757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | ENST00000597757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | ENST00000597757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | ENST00000597757.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | NR_103535.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100014659.hg.1 | NR_103535.1 | Refseq |
| Human Genome U133 Plus 2.0 Array | 1560746_PM_at | - | - |
| Human Genome U133 Plus 2.0 Array | 1560746_at | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560746_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560746_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560746_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560746_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560746_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560746_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560746_at6 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560746_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560746_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560746_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 1560746_at11 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1723567 | AK075118.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1723567 | AK075118.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1723567 | AK075118.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1723567 | AK075118.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1723567 | AK075118.1 | human_transcriptome |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1723567 | AK075118.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1723567 | AK075118.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1723567 | AK075118.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1723567 | AK075118.1 | human_transcriptome |

NEXN-AS1
Genomic Map