Assay Details
| Gene Symbol: TDGF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001174136.1 | NP_001167607.1 | 4 - 5 | 337 | 121 | |
| NM_003212.3 | NP_003203.1 | 4 - 5 | 727 | 121 | ||
| GenBank mRNA | AK313319.1 | - | 4 - 5 | 463 | 121 | |
| BC067844.1 | - | 4 - 5 | 525 | 5286020 | 121 | |
| CX755454.1 | - | 4 - 5 | 336 | 121 | ||
| CX781482.1 | - | 3 - 4 | 298 | 121 | ||
| X14253.1 | - | 4 - 5 | 590 | 121 | ||
| Gene Symbol: TDGF1P3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002718.2 | 1 | 1161 | 121 | ||
| GenBank mRNA | M96956.1 | - | 1 | 1152 | 121 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03003403.hg.1, TC0X000542.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
morphogenesis of a branching structure
cell migration involved in sprouting angiogenesis
epidermal growth factor receptor signaling pathway
heart development
positive regulation of cell proliferation
anterior/posterior axis specification, embryo
embryo development
regulation of signal transduction
positive regulation of endothelial cell migration
peptidyl-serine phosphorylation
cell differentiation
positive regulation of cell migration
mammary gland development
somatic stem cell population maintenance
cellular response to hepatocyte growth factor stimulus
negative regulation of apoptotic process
cellular response to fibroblast growth factor stimulus
positive regulation of peptidyl-tyrosine phosphorylation
cellular response to interferon-gamma
cellular response to interleukin-6
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
signal transduction
receptor binding
protein binding
growth factor activity
activation of MAPK activity
morphogenesis of a branching structure
cell migration involved in sprouting angiogenesis
epidermal growth factor receptor signaling pathway
heart development
positive regulation of cell proliferation
anterior/posterior axis specification, embryo
embryo development
regulation of signal transduction
positive regulation of endothelial cell migration
peptidyl-serine phosphorylation
cell differentiation
positive regulation of cell migration
mammary gland development
somatic stem cell population maintenance
cellular response to hepatocyte growth factor stimulus
negative regulation of apoptotic process
cellular response to fibroblast growth factor stimulus
positive regulation of peptidyl-tyrosine phosphorylation
cellular response to interferon-gamma
cellular response to interleukin-6
cellular response to tumor necrosis factor
cellular response to epidermal growth factor stimulus
signal transduction
receptor binding
protein binding
growth factor activity
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000459867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000459867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000459867.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000471721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000471721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000471721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000471721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000471721.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007267.hg.1 | TC0X000542.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR0X007267.hg.1 | NR_002718.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005260.hg.1 | TC03003403.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005260.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005260.hg.1 | ENST00000542931.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005260.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03005260.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008121.hg.1 | ENST00000493351.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008121.hg.1 | ENST00000602699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008121.hg.1 | ENST00000602699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008121.hg.1 | ENST00000602699.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008121.hg.1 | NR_002718.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008121.hg.1 | NR_002718.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00008121.hg.1 | NR_002718.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000459867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000459867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000459867.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000471721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000471721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000471721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000471721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000471721.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000493282.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300013824.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300204037.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300204037.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300204037.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300204037.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300204037.hg.1 | ENST00000296145.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300204037.hg.1 | ENST00000542931.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300204037.hg.1 | NM_001174136.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300204037.hg.1 | NM_003212.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300204037.hg.1 | TC0300013824.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00155528.hg.1 | ENST00000493351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155528.hg.1 | ENST00000602699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155528.hg.1 | NR_002718.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155528.hg.1 | ENST00000493351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155528.hg.1 | ENST00000602699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155528.hg.1 | NR_002718.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155528.hg.1 | TC0X00008121.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0X00008121.hg.1 | ENST00000493351.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008121.hg.1 | ENST00000602699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008121.hg.1 | ENST00000602699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008121.hg.1 | ENST00000602699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00008121.hg.1 | NR_002718.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008121.hg.1 | NR_002718.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00008121.hg.1 | NR_002718.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155529.hg.1 | ENST00000602699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155529.hg.1 | NR_002718.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155529.hg.1 | ENST00000602699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155529.hg.1 | NR_002718.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155529.hg.1 | TC0X00008121.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0X00155527.hg.1 | ENST00000602699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155527.hg.1 | NR_002718.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155527.hg.1 | ENST00000602699.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0X00155527.hg.1 | NR_002718.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0X00155527.hg.1 | TC0X00008121.hg.1 | Clariom D TC |

TDGF1
Genomic Map