Assay Details
| Gene Symbol: EIF2S2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001316363.1 | NP_001303292.1 | 2 - 3 | 440 | 136 | |
| NM_001316364.1 | NP_001303293.1 | 2 - 3 | 440 | 136 | ||
| NM_003908.4 | NP_003899.2 | 2 - 3 | 440 | 136 | ||
| XM_017028118.1 | XP_016883607.1 | 2 - 3 | 602 | 136 | ||
| GenBank mRNA | AB451237.1 | - | 1 - 2 | 194 | 136 | |
| AB451362.1 | - | 1 - 2 | 194 | 136 | ||
| AK313025.1 | - | 2 - 3 | 311 | 136 | ||
| BC000461.2 | - | 2 - 3 | 330 | 2822324 | 136 | |
| BC000934.2 | - | 2 - 3 | 305 | 3447202 | 136 | |
| BC007888.2 | - | 1 - 2 | 121 | 4120898 | 136 | |
| BT009795.1 | - | 1 - 2 | 194 | 136 | ||
| BX418731.2 | - | 2 - 3 | 310 | 136 | ||
| BX429051.2 | - | 2 - 3 | 286 | 136 | ||
| CR456734.1 | - | 1 - 2 | 194 | 136 | ||
| M29536.1 | - | 2 - 3 | 310 | 136 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC007888.2 transcripts.
Set Membership: |
|

EIF2S2
Genomic Map