Assay Details
| Gene Symbol: NCR3LG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001202439.2 | NP_001189368.1 | 3 - 4 | 965 | 102 | |
| XM_011520072.2 | XP_011518374.1 | 3 - 4 | 2919 | 102 | ||
| XM_011520073.2 | XP_011518375.1 | 3 - 4 | 2665 | 102 | ||
| XM_011520074.2 | XP_011518376.1 | 3 - 4 | 1282 | 102 | ||
| XM_011520075.2 | XP_011518377.1 | 3 - 4 | 2571 | 102 | ||
| GenBank mRNA | BC136797.1 | - | 3 - 4 | 828 | 102 | |
| BC136800.1 | - | 3 - 4 | 828 | 102 | ||
| BX537685.1 | - | 3 - 4 | 957 | 102 | ||
| CR749521.1 | - | 3 - 4 | 965 | 102 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC11000229.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066810 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066868 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667181 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066854 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16424019 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667167 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066855 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667179 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066812 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667160 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667199 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066833 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667201 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667193 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066839 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667144 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667147 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667164 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667175 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667194 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066824 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066826 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066850 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667151 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066837 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667155 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667187 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16424023 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667148 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066849 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667177 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667156 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667180 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066808 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667207 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667154 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667188 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066806 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066835 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667191 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066807 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667183 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667158 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066805 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667186 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667157 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667153 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667162 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667178 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066867 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667197 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667169 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28066869 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07667182 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3356210 | - | - |
| Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 72601 | - | - |
| Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 82903 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19165606 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19165598 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02401658 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P39281641 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P19165605 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P02401646 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 7938703 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7938705 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7938706 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7938704 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7938702 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003007.hg.1 | ENST00000338965.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003007.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR11003007.hg.1 | TC11000229.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1100006966.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149031.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149031.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149031.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149031.hg.1 | ENST00000338965.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149031.hg.1 | ENST00000530403.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1100149031.hg.1 | NM_001202439.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1100149031.hg.1 | TC1100006966.hg.1 | Clariom D TC |
| Human Genome U95 Set | 46881_PM_at | - | - |
| Human Genome U95 Set | 46881_at | - | - |
| Human Genome U95 Set | 43752_at | - | - |
| Human Genome U95 Set | 43752_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046181 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046181 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1669972 | BX537685.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1669972 | CR749521.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1669972 | NM_001202439.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1669972 | BX537685.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1669972 | CR749521.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1669972 | NM_001202439.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1669972 | BX537685.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1669972 | CR749521.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1669972 | NM_001202439.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1669972 | BX537685.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1669972 | CR749521.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1669972 | NM_001202439.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1669972 | BX537685.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1669972 | CR749521.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1669972 | NM_001202439.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1669972 | BX537685.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1669972 | CR749521.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1669972 | NM_001202439.1 | human_transcriptome |

NCR3LG1
Genomic Map