Assay Details
| Gene Symbol: TPM3P9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003148.3 | 2 | 940 | 144 | ||
| GenBank mRNA | AK094777.1 | - | 4 | 880 | 144 | |
| BC061910.1 | - | 2 | 751 | 144 | ||
| BX647962.1 | - | 2 | 927 | 144 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10997716 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P17158859 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10997719 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P10997717 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3216433 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_23_P55846 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 31480 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 17570 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3216433 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3216433 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17158864 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10997720 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10997717 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31638937 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10997722 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10997719 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17158859 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31638945 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10997718 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17158861 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10997721 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10997719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17158864 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17158861 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17158859 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31638937 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10997720 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10997718 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17158864 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10997722 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17158861 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10997719 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P31638945 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P17158859 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10997721 | - | - |
| Agilent Whole Chr array_18 19 21 | A_17_P10997717 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P10997719 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17158861 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17158859 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P17158864 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_23_P55846 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_24_P211206 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P55846 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03463757 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10997721 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10997717 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P10997718 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_003148_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8030992 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8030991 | - | - |
| Human Genome U133 Plus 2.0 Array | 234620_at | - | - |
| Human Genome U133 Plus 2.0 Array | 234620_PM_at | - | - |
| Human Genome U133 Set | 234620_at | - | - |
| Human Genome U133 Set | 234620_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007129 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A007129 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1761801 | CR610971.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1761801 | BC047783.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1761801 | BC061910.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1761801 | BX647962.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_1761801 | NR_003148.3 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1761801 | CR610971.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1761801 | BC047783.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1761801 | BC061910.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1761801 | BX647962.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1761801 | NR_003148.3 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1761801 | CR610971.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1761801 | BC047783.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1761801 | BC061910.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1761801 | BX647962.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1761801 | NR_003148.3 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1761801 | CR610971.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1761801 | BC047783.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1761801 | BC061910.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1761801 | BX647962.1 | human_transcriptome |
| Illumina human-6 v2.0 expression beadchip | ILMN_1761801 | NR_003148.3 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1761801 | CR610971.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1761801 | BC047783.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1761801 | BC061910.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1761801 | BX647962.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1761801 | NR_003148.3 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1761801 | CR610971.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1761801 | BC047783.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1761801 | BC061910.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1761801 | BX647962.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1761801 | NR_003148.3 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1761801 | CR610971.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1761801 | BC047783.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1761801 | BC061910.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1761801 | BX647962.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1761801 | NR_003148.3 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1761801 | CR610971.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1761801 | BC047783.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1761801 | BC061910.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1761801 | BX647962.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1761801 | NR_003148.3 | human_transcriptome |

TPM3P9
Genomic Map