Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0600010359.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170630.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170630.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170630.hg.1 |
ENST00000580119.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0600170630.hg.1 |
NR_126020.1 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0600170630.hg.1 |
TC0600010359.hg.1 |
Clariom D TC |