Assay Details
| Gene Symbol: HLA-F-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AL832418.1 | - | 5 - 6 | 1060 | 112 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000399247.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000428990.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000434086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000434086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000434086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000434086.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000458236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000458236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000458236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000458236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000458236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000458236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000458236.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000510438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000510438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000510438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000510438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000510438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | ENST00000510438.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026972.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026973.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600011323.hg.1 | NR_026973.1 | Refseq |

HLA-F-AS1
Genomic Map