Assay Details
| Gene Symbol: OR4P4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001004124.2 | NP_001004124.1 | 1 | 759 | 111 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
G-protein coupled receptor activity
olfactory receptor activity
G-protein coupled receptor signaling pathway
detection of chemical stimulus involved in sensory perception
detection of chemical stimulus involved in sensory perception of smell
transmembrane signaling receptor activity
G-protein coupled receptor activity
olfactory receptor activity
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215279 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07806156 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07806177 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215302 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215267 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16452422 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07806174 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215297 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16452419 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215278 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07806190 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215270 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215312 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215273 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215282 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215284 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215275 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215296 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215281 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215300 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215274 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215292 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215269 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215309 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215285 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07806164 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215299 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07806150 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16452429 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215271 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215286 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215314 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215313 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215310 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215294 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215266 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215287 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215288 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215308 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07806184 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215304 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215283 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P07806183 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P16452430 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P28215277 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 133469 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3354836 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0285 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0287 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0286 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 3902 | - | - |
| Agilent Human Genome CGH + SNP Microarray | A_16_P32908038 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3354836 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3354836 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806184 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215292 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215288 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215302 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215274 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215266 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215275 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806179 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806156 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16452429 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215309 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16452422 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215300 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215278 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215302 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215288 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215268 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215271 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215286 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215281 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215274 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806184 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215300 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215297 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215304 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215277 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215279 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215282 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806179 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806150 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215284 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215294 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806156 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215287 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215266 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215292 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215299 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215269 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215278 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215267 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P07806183 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215309 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16452429 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P16452422 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P28215275 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806164 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806162 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806184 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806169 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806151 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806175 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806159 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806167 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806186 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806177 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806183 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806150 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806170 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806187 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806181 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806157 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806173 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806172 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806153 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806185 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806180 | - | - |
| Agilent Human Promoter ChIP-on-Chip A) | A_17_P07806156 | - | - |
| Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P39361721 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P39361721 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806156 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806211 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806170 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806173 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16452420 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806205 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806159 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806157 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806220 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806169 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806177 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806201 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806162 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16452426 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806150 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16452429 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806215 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806185 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806152 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806172 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806192 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806181 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16452422 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806187 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806217 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806164 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P16452431 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806180 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806223 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806206 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806203 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806175 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806183 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806167 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806208 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P07806190 | - | - |
| Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P39361721 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NM_001004124_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7939929 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 7939928 | - | - |
| Affymetrix Human Genome U219 Array | 11742602_PM_at | - | - |
| Affymetrix Human Genome U219 Array | 11742602_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046379 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A046379 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_1804845 | NM_001004124.1 | human_transcriptome |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1804845 | NM_001004124.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_1804845 | NM_001004124.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1804845 | NM_001004124.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1804845 | NM_001004124.1 | human_transcriptome |
| Illumina HumanRef-8 v2 Expression BeadChip | GI_3498-S | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1804845 | NM_001004124.1 | human_transcriptome |
| Illumina humanRef-8 v2.0 expression beadchip | ILMN_1804845 | NM_001004124.1 | human_transcriptome |
| Illumina Sentrix Human-6 v2 Expression BeadChip | 2600180 | - | - |
| Illumina HumanRef-6 v2.0 expression beadchip | ILMN_3498 | - | - |
| Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1804845 | NM_001004124.1 | human_transcriptome |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1804845 | NM_001004124.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1804845 | NM_001004124.1 | human_transcriptome |
| Illumina HumanWG-6_V2_0_R2 | ILMN_1804845 | NM_001004124.1 | human_transcriptome |
| Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1804845 | NM_001004124.1 | human_transcriptome |

OR4P4
Genomic Map