Assay Details
| Gene Symbol: NAA30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001011713.2 | NP_001011713.2 | 2 - 3 | 920 | 150 | |
| GenBank mRNA | AK092674.1 | - | 2 - 3 | 920 | 150 | |
| AK304257.1 | - | 2 - 3 | 524 | 150 | ||
| BC118589.1 | - | 2 - 3 | 875 | 40085064 | 150 | |
| BC122557.1 | - | 3 - 4 | 764 | 40085054 | 150 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14000342.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14003652.hg.1 | ENST00000395257.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003652.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003652.hg.1 | ENST00000555166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003652.hg.1 | ENST00000556492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003652.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003652.hg.1 | TC14000342.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003649.hg.1 | ENST00000395257.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003649.hg.1 | ENST00000556492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003649.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003649.hg.1 | TC14000342.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003650.hg.1 | ENST00000556492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003650.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003650.hg.1 | TC14000342.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003645.hg.1 | ENST00000555166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003645.hg.1 | ENST00000556492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003645.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003645.hg.1 | TC14000342.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003647.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003647.hg.1 | ENST00000555166.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003647.hg.1 | ENST00000556492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003647.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003647.hg.1 | TC14000342.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003651.hg.1 | ENST00000395257.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003651.hg.1 | ENST00000556492.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003651.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14003651.hg.1 | TC14000342.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1400150483.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150483.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150483.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150483.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150483.hg.1 | TC1400007285.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150484.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150484.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150484.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150484.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150484.hg.1 | TC1400007285.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150482.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150482.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150482.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150482.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150482.hg.1 | TC1400007285.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150487.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150487.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150487.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150487.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150487.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150487.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150487.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150487.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150487.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150487.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150487.hg.1 | TC1400007285.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150486.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150486.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150486.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150486.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150486.hg.1 | TC1400007285.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150478.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150478.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150478.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150478.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150478.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150478.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150478.hg.1 | TC1400007285.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150480.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150480.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150480.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150480.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150480.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150480.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150480.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150480.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150480.hg.1 | TC1400007285.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1400150485.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150485.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150485.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150485.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150485.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400150485.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400150485.hg.1 | TC1400007285.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000298406.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000554703.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000555166.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | ENST00000556492.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400007285.hg.1 | NM_001011713.2 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 124900 | - | - |

NAA30
Genomic Map