Assay Details
| Gene Symbol: FAM86B2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001137610.1 | NP_001131082.1 | 7 - 8 | 897 | 108 | |
| XM_005273603.4 | XP_005273660.1 | 6 - 7 | 870 | 108 | ||
| XM_005273604.4 | XP_005273661.1 | 5 - 6 | 789 | 108 | ||
| XM_005273605.4 | XP_005273662.1 | 5 - 6 | 690 | 108 | ||
| XM_006716387.3 | XP_006716450.1 | 6 - 7 | 891 | 108 | ||
| XM_011544618.2 | XP_011542920.1 | 7 - 8 | 942 | 108 | ||
| XM_011544619.2 | XP_011542921.1 | 6 - 7 | 909 | 108 | ||
| XM_011544622.2 | XP_011542924.1 | 6 - 7 | 811 | 108 | ||
| XM_017013740.1 | XP_016869229.1 | 5 - 6 | 872 | 108 | ||
| XM_017013741.1 | XP_016869230.1 | 4 - 5 | 809 | 108 | ||
| XM_017013742.1 | XP_016869231.1 | 4 - 5 | 791 | 108 | ||
| XM_017013743.1 | XP_016869232.1 | 4 - 5 | 491 | 108 | ||
| GenBank mRNA | AK297741.1 | - | 4 - 5 | 472 | 108 | |
| Gene Symbol: FAM86B1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK098138.1 | - | 5 - 6 | 541 | 108 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000995.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3310430 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P280868 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3269740 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 36385 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 55677 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 27959 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15429 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3269740 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3310430 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_24_P280868 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3269740 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_24_P280868 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_33_P3310430 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_24_P280868 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8149375 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149393 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149394 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149374 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149369 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149373 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149391 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149390 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149372 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149392 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149370 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149376 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149395 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8149371 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 7945877 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8149389 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8149365 | - | - |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000515046.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000515046.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000515046.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000515046.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012424.hg.1 | TC08000995.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012424.hg.1 | ENST00000262365.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012424.hg.1 | ENST00000309608.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012424.hg.1 | ENST00000351291.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012424.hg.1 | ENST00000393715.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012424.hg.1 | NM_001137610.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012426.hg.1 | TC08000995.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012426.hg.1 | ENST00000262365.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012426.hg.1 | ENST00000309608.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012426.hg.1 | ENST00000351291.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012426.hg.1 | ENST00000393715.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08012426.hg.1 | NM_001137610.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165616.hg.1 | ENST00000262365.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165616.hg.1 | NM_001137610.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165616.hg.1 | ENST00000262365.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165616.hg.1 | NM_001137610.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165616.hg.1 | TC0800009656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800165619.hg.1 | ENST00000262365.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165619.hg.1 | ENST00000309608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165619.hg.1 | ENST00000527331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165619.hg.1 | NM_001137610.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165619.hg.1 | ENST00000262365.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165619.hg.1 | ENST00000309608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165619.hg.1 | ENST00000527331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165619.hg.1 | NM_001137610.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165619.hg.1 | TC0800009656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800165617.hg.1 | ENST00000262365.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165617.hg.1 | ENST00000309608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165617.hg.1 | NM_001137610.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165617.hg.1 | ENST00000262365.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165617.hg.1 | ENST00000309608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165617.hg.1 | NM_001137610.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165617.hg.1 | TC0800009656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800165620.hg.1 | ENST00000262365.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165620.hg.1 | ENST00000309608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165620.hg.1 | ENST00000527331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165620.hg.1 | NM_001137610.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165620.hg.1 | ENST00000262365.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165620.hg.1 | ENST00000309608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165620.hg.1 | ENST00000527331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165620.hg.1 | NM_001137610.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165620.hg.1 | TC0800009656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800165621.hg.1 | ENST00000262365.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165621.hg.1 | ENST00000309608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165621.hg.1 | ENST00000527331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165621.hg.1 | ENST00000532480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165621.hg.1 | NM_001137610.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165621.hg.1 | ENST00000262365.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165621.hg.1 | ENST00000309608.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165621.hg.1 | ENST00000527331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165621.hg.1 | ENST00000532480.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800165621.hg.1 | NM_001137610.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800165621.hg.1 | TC0800009656.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000433621.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000515046.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000515046.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000515046.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000515046.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000525829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | ENST00000602658.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012272.hg.1 | NR_026789.1 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025278 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035538 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035532 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035532 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A035538 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A025278 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3262462 | NM_001137610.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3262462 | BE729618.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3262462 | DA552021.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3262462 | NM_001083537.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3186004 | - | - |
| Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1677069 | - | - |
| Illumina human-6 v2.0 expression beadchip | ILMN_1677069 | - | - |
| Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1806633 | AK298940.1 | human_transcriptome |
| Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1806633 | AK298940.1 | human_transcriptome |

FAM86B2
Genomic Map