Assay Details
| Gene Symbol: TCEAL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006612.1 | NP_001006613.1 | 3 | 458 | 94 | |
| NM_001006613.1 | NP_001006614.1 | 2 | 382 | 94 | ||
| NM_001006614.1 | NP_001006615.1 | 2 | 379 | 94 | ||
| NM_016303.2 | NP_057387.1 | 3 | 461 | 94 | ||
| GenBank mRNA | AF125535.1 | - | 3 | 419 | 94 | |
| AL833152.1 | - | 1 | 1325 | 94 | ||
| BC023544.2 | - | 3 | 412 | 4122244 | 94 | |
| BG400091.1 | - | 3 | 462 | 94 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TCEAL9
Genomic Map