| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014187.hg.1 |
TC0X001057.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014187.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014187.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014186.hg.1 |
TC0X001057.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014186.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014186.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014185.hg.1 |
TC0X001057.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014185.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X014185.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009727.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009727.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009727.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009727.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009727.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009727.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00009727.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166440.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166440.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166440.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166440.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166440.hg.1 |
TC0X00009727.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00166437.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166437.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166437.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166437.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166437.hg.1 |
TC0X00009727.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00166439.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166439.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166439.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00166439.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00166439.hg.1 |
TC0X00009727.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00009727.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009727.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009727.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009727.hg.1 |
ENST00000375511.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00009727.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009727.hg.1 |
NM_001009616.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00009727.hg.1 |
NM_001009616.3 |
Refseq |