Assay Details
| Gene Symbol: PMS2P6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AB017005.1 | - | 8 | 861 | 175 | |
More Information
Set Membership: |
|
Panther Classification:
MicroArray Probes:
Agilent
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 779597 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 557770 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 344482 | - | - |
| Agilent SurePrint G3 Human CNV Microarray 2x400K | 188006 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | 8133302 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8133311 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140277 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140246 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140276 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8133304 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8133312 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140275 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140244 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140263 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8133303 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140272 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140261 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140259 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140262 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140273 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140266 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140264 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140274 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140245 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8133310 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140241 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140265 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140270 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140243 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8140242 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8140239 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8133309 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8133300 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8140258 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8140269 | - | - |
| AffymetrixPathwork Diagnostics Pathchip | pw528969 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215410_at4 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215410_at5 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215410_at1 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215410_at9 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215410_at10 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215410_at2 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215410_at11 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215410_at7 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215410_at3 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215410_at8 | - | - |
| Affymetrix Human Genome U133 Plus 2.0 Array | 215410_at6 | - | - |

PMS2P6
Genomic Map