| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008944.hg.1 |
TC0X000694.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008944.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008944.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008944.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008942.hg.1 |
TC0X000694.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008942.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008942.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X008942.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008621.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008621.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008621.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008621.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008621.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008621.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008621.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008621.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008621.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008621.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008621.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0X00008621.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158532.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158532.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158532.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158532.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158532.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158532.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158532.hg.1 |
TC0X00008621.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0X00158534.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158534.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158534.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158534.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158534.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158534.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158534.hg.1 |
TC0X00008621.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0X00008621.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008621.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008621.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008621.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008621.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008621.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008621.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008621.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0X00008621.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008621.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008621.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0X00008621.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158531.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158531.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158531.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158531.hg.1 |
ENST00000370504.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158531.hg.1 |
ENST00000446864.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00158531.hg.1 |
NM_001009613.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00158531.hg.1 |
TC0X00008621.hg.1 |
Clariom D TC |