Assay Details
| Gene Symbol: GABRR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105580.2 | NP_001099050.1 | 2 - 3 | 243 | 73 | |
| NR_047685.1 | 2 - 3 | 243 | 73 | |||
| GenBank mRNA | CR749263.1 | - | 2 - 3 | 252 | 73 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001600.hg.1
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR03028791.hg.1 | TC03001600.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028791.hg.1 | ENST00000472788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028791.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028792.hg.1 | TC03001600.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028792.hg.1 | ENST00000472788.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028792.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028793.hg.1 | TC03001600.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR03028793.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187639.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187639.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187639.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187639.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187639.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187639.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187639.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187639.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187639.hg.1 | TC0300011802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300187642.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187642.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187642.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187642.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187642.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187642.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187642.hg.1 | TC0300011802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300187640.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187640.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187640.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187640.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187640.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187640.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187640.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300187640.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300187640.hg.1 | TC0300011802.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |

GABRR3
Genomic Map