Assay Details
| Gene Symbol: GABRR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | CR749263.1 | - | 5 - 6 | 507 | 142 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000470589.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000472788.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | ENST00000621172.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300011802.hg.1 | NM_001105580.2 | Refseq |

GABRR3
Genomic Map