Assay Details
| Gene Symbol: CA5BP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC021816.1 | - | 1 - 3 | 38 | 65 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380331.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380333.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380333.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380333.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380333.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380333.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380333.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000448692.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000448692.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000448692.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000448692.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | ENST00000448692.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380331.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380333.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380333.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380333.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380333.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380333.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380333.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380334.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000380336.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000435806.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000448692.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000448692.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000448692.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000448692.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | ENST00000448692.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | NR_026551.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0X00011276.hg.1 | NR_026551.1 | Refseq |

CA5BP1
Genomic Map