Assay Details
| Gene Symbol: FAM92A1P2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003612.2 | 1 | 2259 | 86 | ||
| GenBank mRNA | BC022019.1 | - | 1 | 2222 | 86 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676414 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837381 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676450 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837335 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837354 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676439 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837367 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837389 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837382 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837387 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837370 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676425 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837386 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676446 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676422 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676438 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676443 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837349 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676454 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837347 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837346 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676442 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837364 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837333 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837361 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837373 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837351 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837371 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676445 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837338 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676453 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676417 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837355 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837356 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837350 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P03676437 | - | - |
| Agilent Human Whole Genome ChIP Tiling Array | A_17_P23837385 | - | - |
| Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 272254 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P94001 | - | - |
| Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_44k_32_P94001 | - | - |
| Agilent Whole Human Genome Oligo Microarray 4x44K | 9320 | - | - |
| Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 48817 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P103695 | - | - |
| Agilent Whole Human Genome Microarray 4x44K v2 | A_32_P94001 | - | - |
| SurePrint G3 Human GE Microarray G4851A | A_32_P94001 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676422 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837382 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837384 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837346 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676414 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676420 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676421 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837387 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837357 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676443 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676425 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837338 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676450 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837356 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837350 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837343 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15644652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837349 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676451 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837373 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676456 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676417 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676445 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837351 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837371 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676438 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837385 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837367 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676456 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676417 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676421 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676451 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837387 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676443 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837384 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837370 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837373 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837382 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837349 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837343 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837361 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676438 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676445 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676422 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837354 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837356 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676450 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676425 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837371 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837350 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837346 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837338 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676420 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676454 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837335 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837351 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837386 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837357 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P23837364 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15644652 | - | - |
| Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P03676414 | - | - |
| Whole Human Genome Oligo Microarray G4112A | A_32_P94001 | - | - |
| Agilent Whole Human Genome Microarray 4x44K G4112F | A_32_P94001 | - | - |
| Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P31416152 | - | - |
| Agilent Human 1B Microarray G4111A | A_32_P94001 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676472 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676455 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676415 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676432 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676463 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676417 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676414 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676440 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676435 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676427 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676451 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676421 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676422 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676465 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676474 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676424 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676429 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676443 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676467 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676430 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676470 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676462 | - | - |
| Agilent Human Whole Genome ChIP-on-Chip | A_17_P03676460 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Gene 1.0 ST Array | NR_003612_at | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8098469 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8098467 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8098466 | - | - |
| Affymetrix Human Gene 1.0 ST Array | 8098468 | - | - |
| Affymetrix Human Gene 1.1 ST Array | 8098465 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026352 | - | - |
| Arraystar Human LncRNA microarray V2.0 | ASHG19A3A026352 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 169180 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| HumanHT-12 v4 Expression BeadChip | ILMN_3235997 | BC022019.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3235997 | BC047622.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3235997 | BC059396.1 | human_transcriptome |
| HumanHT-12 v4 Expression BeadChip | ILMN_3235997 | NR_003612.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3235997 | BC022019.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3235997 | BC047622.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3235997 | BC059396.1 | human_transcriptome |
| HumanHT-12 V4.0 expression beadchip | ILMN_3235997 | NR_003612.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3235997 | BC022019.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3235997 | BC047622.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3235997 | BC059396.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_3235997 | NR_003612.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3235997 | BC022019.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3235997 | BC047622.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3235997 | BC059396.1 | human_transcriptome |
| Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_3235997 | NR_003612.1 | human_transcriptome |

FAM92A1P2
Genomic Map