Assay Details
| Gene Symbol: LOC387720 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK127642.1 | - | 1 | 2252 | 158 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | neuropeptide S | NPS | -181.9kb | Anti-sense | Chr. 10 - 127549349 - 127552671 on NCBI Build GRCh38 | |
| Downstream | forkhead box I2 | FOXI2 | 278.0 | Anti-sense | Chr. 10 - 127737274 - 127741186 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LOC387720
Genomic Map