Assay Details
Gene Symbol: LINC00602 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_027284.1 | 1 | 2001 | 94 | ||
GenBank mRNA | AK094116.1 | - | 1 | 2001 | 94 | |
BC110806.1 | - | 1 | 1925 | 94 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239676 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239669 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239634 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25444679 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239674 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239668 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15887997 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239681 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239661 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239635 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15887981 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25444694 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25444692 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239678 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25444699 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25444688 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239667 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239654 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239655 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239677 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15887998 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25444701 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239682 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239648 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25444693 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25444653 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25444687 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25444676 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239649 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239636 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15887982 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P05239647 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P25444682 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P326414 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5097 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_5098 | - | - |
Agilent Whole Human Genome Oligo Microarray 4x44K | 18865 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 53792 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_23_P326414 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_23_P326414 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444584 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239600 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444653 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444687 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444597 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444615 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444691 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239648 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239622 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444611 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239624 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239585 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239634 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444625 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444633 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239589 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444594 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239655 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15887980 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239607 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444612 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239571 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239575 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444607 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239573 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444693 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444695 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444580 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444644 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15887982 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15887998 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239565 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444599 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239636 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239572 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239647 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239632 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444586 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444601 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239604 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444692 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239594 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239560 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239618 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239603 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444619 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444628 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444605 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239649 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444623 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239635 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239604 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239600 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444692 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444687 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444688 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444601 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444678 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15887998 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239655 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239626 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239603 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239632 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239589 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444682 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444680 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239575 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444653 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239635 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444612 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444695 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444625 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239636 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444694 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15887982 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239622 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444605 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444586 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239571 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239560 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239585 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444597 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239648 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239573 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239634 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444693 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239667 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444599 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239594 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239624 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239647 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444592 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444615 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239618 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444623 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444691 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444628 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444584 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444633 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239629 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444580 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444644 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239565 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444619 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444676 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444594 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239607 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444607 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15887980 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239649 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P05239572 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P25444611 | - | - |
Whole Human Genome Oligo Microarray G4112A | A_23_P326414 | - | - |
Agilent Whole Human Genome Microarray 4x44K G4112F | A_23_P326414 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239678 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239636 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239618 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239630 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239670 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239679 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239665 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239576 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239593 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239557 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239672 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239585 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239649 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239667 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239562 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239632 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239570 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239676 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239595 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15887999 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239572 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239647 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239622 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15887978 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239601 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239603 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239583 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239681 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239609 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239616 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239587 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239589 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239653 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239559 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239658 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239620 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239655 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239578 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239573 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239629 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239567 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239612 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239648 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239596 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239634 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239637 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239624 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239674 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239581 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239597 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239606 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15887997 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P05239626 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15887998 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8123335 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8123334 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A032314 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A032314 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1742967 | AK094116.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1742967 | BC110806.1 | human_transcriptome |
HumanHT-12 v4 Expression BeadChip | ILMN_1742967 | NR_027284.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1792308 | - | - |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1742967 | AK094116.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1742967 | BC110806.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1742967 | NR_027284.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1792308 | - | - |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1792308 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1742967 | AK094116.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1742967 | BC110806.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1742967 | NR_027284.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1742967 | AK094116.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1742967 | BC110806.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1742967 | NR_027284.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1742967 | AK094116.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1742967 | BC110806.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1742967 | NR_027284.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1742967 | AK094116.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1742967 | BC110806.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1742967 | NR_027284.1 | human_transcriptome |
