Assay Details
| Gene Symbol: CCNO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125346.1 | 1 - 2 | 957 | 58 | ||
| GenBank mRNA | BC004877.2 | - | 1 - 2 | 837 | 58 | |
| BC143907.1 | - | 1 - 2 | 112 | 58 | ||
More Information
Updated Mapping Information
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500010759.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175046.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175046.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175046.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175046.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175046.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175046.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175046.hg.1 | TC0500010759.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500010759.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175044.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175044.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175044.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175044.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175044.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175044.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175044.hg.1 | ENST00000282572.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175044.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175044.hg.1 | NM_021147.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175044.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175044.hg.1 | NR_125347.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175044.hg.1 | NR_125348.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175044.hg.1 | TC0500010759.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0500175047.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175047.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175047.hg.1 | ENST00000501463.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500175047.hg.1 | NR_125346.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500175047.hg.1 | TC0500010759.hg.1 | Clariom D TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 169995 | - | - |

CCNO
Genomic Map