Assay Details
| Gene Symbol: SLC7A5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003486.5 | NP_003477.4 | 10 | 4296 | 101 | |
| XM_006721286.3 | XP_006721349.2 | 10 | 3939 | 101 | ||
| GenBank mRNA | AB018009.1 | - | 10 | 4291 | 101 | |
| AF104032.1 | - | 10 | 4293 | 101 | ||
| BC014177.2 | - | 1 | 1952 | 101 | ||
| BC039692.2 | - | 10 | 4322 | 5551612 | 101 | |
| BC042600.1 | - | 10 | 4292 | 4470593 | 101 | |
| M80244.1 | - | 10 | 3736 | 101 | ||
More Information
Updated Mapping Information
Set Membership: |
|

SLC7A5
Genomic Map