Assay Details
| Gene Symbol: ABCC12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033226.2 | NP_150229.2 | 20 | 3131 | 79 | |
| GenBank mRNA | AF395908.1 | - | 20 | 2782 | 79 | |
| AF411577.1 | - | 20 | 2782 | 79 | ||
| AF411578.1 | - | 20 | 2782 | 79 | ||
| AY040220.1 | - | 20 | 3131 | 79 | ||
| AY196326.1 | - | 18 | 2765 | 79 | ||
| BC036378.1 | - | 21 | 3207 | 79 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

ABCC12
Genomic Map