Assay Details
| Gene Symbol: SNX29 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011522743.2 | XP_011521045.1 | 22 | 3369 | 59 | |
| XM_011522744.2 | XP_011521046.1 | 21 | 3129 | 59 | ||
| GenBank mRNA | BC029857.1 | - | 9 | 1492 | 59 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

SNX29
Genomic Map