Assay Details
Gene Symbol: UGT2B7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001074.2 | NP_001065.2 | 1 | 348 | 113 | |
XM_011532229.2 | XP_011530531.1 | 1 | 349 | 113 | ||
XM_011532230.1 | XP_011530532.1 | 1 | 349 | 113 | ||
GenBank mRNA | AK223142.1 | - | 1 | 322 | 113 | |
AK313190.1 | - | 1 | 349 | 113 | ||
BC030974.1 | - | 1 | 348 | 4593426 | 113 | |
J05428.1 | - | 1 | 316 | 113 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000371.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | PSR04005027.hg.1 | TC04000371.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR04005027.hg.1 | ENST00000305231.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005027.hg.1 | ENST00000508661.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR04005027.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0400007729.hg.1 | ENST00000509466.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0400007729.hg.1 | ENST00000509466.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000502942.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000509763.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | TC0400007749.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400152791.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152791.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152791.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152791.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400152791.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152791.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152791.hg.1 | ENST00000622664.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152791.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400152791.hg.1 | TC0400007749.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0400007729.hg.1 | ENST00000509466.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0400007729.hg.1 | ENST00000509466.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152789.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152789.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152789.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400152789.hg.1 | ENST00000305231.11 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152789.hg.1 | ENST00000508661.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0400152789.hg.1 | NM_001074.3 | Refseq |
Affymetrix Human Clariom D Assay | PSR0400152789.hg.1 | TC0400007749.hg.1 | Clariom D TC |
