Assay Details
| Gene Symbol: GSK3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001146156.1 | NP_001139628.1 | 5 | 1517 | 78 | |
| NM_002093.3 | NP_002084.2 | 5 | 1517 | 78 | ||
| XM_006713610.2 | XP_006713673.1 | 5 | 1003 | 78 | ||
| XM_006713611.2 | XP_006713674.1 | 5 | 1003 | 78 | ||
| GenBank mRNA | AB451356.1 | - | 5 | 534 | 78 | |
| AK290897.1 | - | 5 | 890 | 78 | ||
| BC000251.1 | - | 5 | 766 | 3357620 | 78 | |
| BC012760.2 | - | 5 | 1516 | 3637163 | 78 | |
| CR536510.1 | - | 5 | 534 | 78 | ||
| EU302497.1 | - | 5 | 534 | 78 | ||
| L33801.1 | - | 5 | 573 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

GSK3B
Genomic Map