Assay Details
| Gene Symbol: WRN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000553.4 | NP_000544.2 | 9 - 10 | 2059 | 107 | |
| XM_011544639.2 | XP_011542941.1 | 9 - 10 | 1541 | 107 | ||
| GenBank mRNA | AF091214.1 | - | 9 - 10 | 1502 | 107 | |
| AY818673.1 | - | 8 - 9 | 1271 | 107 | ||
More Information
Set Membership: |
|

WRN
Genomic Map