Assay Details
| Gene Symbol: HRNR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009931.2 | NP_001009931.1 | 1 - 2 | 54 | 94 | |
| GenBank mRNA | AB180729.1 | - | 1 - 2 | 51 | 92 | |
| AY396741.1 | - | 1 - 2 | 54 | 94 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB180729.1, AY396741.1, NM_001009931.2 transcripts.
Set Membership: |
|

HRNR
Genomic Map