Assay Details
| Gene Symbol: TLL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_012464.4 | NP_036596.3 | 21 | 6273 | 157 | |
| XM_011532214.1 | XP_011530516.1 | 21 | 6331 | 157 | ||
| XM_017008570.1 | XP_016864059.1 | 22 | 6399 | 157 | ||
| GenBank mRNA | AK021908.1 | - | 1 | 1306 | 157 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000833.hg.1
Set Membership: |
|

TLL1
Genomic Map