Assay Details
| Gene Symbol: DHX30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014966.3 | NP_055781.2 | 16 | 2822 | 88 | |
| NM_138615.2 | NP_619520.1 | 15 | 2793 | 88 | ||
| XM_006713033.1 | XP_006713096.1 | 14 | 2616 | 88 | ||
| XM_011533490.1 | XP_011531792.1 | 17 | 3048 | 88 | ||
| XM_011533491.1 | XP_011531793.1 | 18 | 3235 | 88 | ||
| XM_011533492.2 | XP_011531794.1 | 16 | 3067 | 88 | ||
| XM_011533493.1 | XP_011531795.1 | 16 | 2937 | 88 | ||
| XM_011533494.2 | XP_011531796.1 | 15 | 2757 | 88 | ||
| XM_011533495.1 | XP_011531797.1 | 15 | 2602 | 88 | ||
| XM_011533496.1 | XP_011531798.1 | 16 | 2954 | 88 | ||
| XM_011533497.2 | XP_011531799.1 | 16 | 2835 | 88 | ||
| XM_011533498.1 | XP_011531800.1 | 16 | 2905 | 88 | ||
| XM_017005914.1 | XP_016861403.1 | 16 | 2698 | 88 | ||
| XM_017005915.1 | XP_016861404.1 | 14 | 2573 | 88 | ||
| XM_017005916.1 | XP_016861405.1 | 14 | 2658 | 88 | ||
| XM_017005917.1 | XP_016861406.1 | 17 | 2864 | 88 | ||
| GenBank mRNA | AB020697.1 | - | 15 | 2510 | 88 | |
| AK291266.1 | - | 16 | 2597 | 88 | ||
| BC014237.2 | - | 3 | 288 | 88 | ||
| BC015029.1 | - | 16 | 2568 | 3911049 | 88 | |
| BC020126.1 | - | 15 | 2541 | 4339419 | 88 | |
| BC038417.1 | - | 16 | 2569 | 5171702 | 88 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

DHX30
Genomic Map