Assay Details
| Gene Symbol: MELTF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033316.3 | NP_201573.1 | 7 | 1486 | 136 | |
| GenBank mRNA | BC001875.1 | - | 7 | 1455 | 136 | |
| BC002623.2 | - | 7 | 1441 | 136 | ||
| BC007550.1 | - | 7 | 1455 | 136 | ||
| BC071910.1 | - | 7 | 1447 | 136 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

MELTF
Genomic Map