Assay Details
| Gene Symbol: PUSL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_153339.1 | NP_699170.1 | 7 | 837 | 67 | |
| GenBank mRNA | AK027721.1 | - | 3 | 1682 | 67 | |
| AK075292.1 | - | 7 | 854 | 67 | ||
| AK298218.1 | - | 6 | 747 | 67 | ||
| BC034304.1 | - | 7 | 837 | 4778177 | 67 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000033.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000421.hg.1 | TC01000033.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000421.hg.1 | ENST00000379031.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000421.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01000421.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000463758.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000470520.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100006499.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144746.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144746.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144746.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144746.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144746.hg.1 | ENST00000379031.9 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144746.hg.1 | ENST00000467712.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144746.hg.1 | ENST00000493657.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100144746.hg.1 | NM_153339.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100144746.hg.1 | TC0100006499.hg.1 | Clariom D TC |

PUSL1
Genomic Map