Assay Details
| Gene Symbol: RNF175 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_173662.2 | NP_775933.1 | 7 | 1064 | 79 | |
| XM_005262938.3 | XP_005262995.1 | 7 | 1801 | 79 | ||
| XM_005262939.3 | XP_005262996.1 | 6 | 866 | 79 | ||
| XM_005262940.4 | XP_005262997.1 | 5 | 1003 | 79 | ||
| XM_011531879.2 | XP_011530181.1 | 7 | 1009 | 79 | ||
| XM_011531881.2 | XP_011530183.1 | 7 | 1148 | 79 | ||
| XM_011531882.2 | XP_011530184.1 | 7 | 1812 | 79 | ||
| XM_011531883.2 | XP_011530185.1 | 7 | 1029 | 79 | ||
| XM_017008047.1 | XP_016863536.1 | 5 | 782 | 79 | ||
| GenBank mRNA | AK091509.1 | - | 6 | 684 | 79 | |
| BC034385.1 | - | 7 | 1065 | 5180678 | 79 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000508967.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000347063.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000503694.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506358.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000506505.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000507512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508248.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000508967.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000509321.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | ENST00000513656.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012171.hg.1 | NM_173662.2 | Refseq |

RNF175
Genomic Map