Assay Details
| Gene Symbol: TNFRSF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243.4 | NP_001234.3 | 6 | 854 | 57 | |
| NM_001281430.2 | NP_001268359.2 | 5 | 766 | 57 | ||
| XM_011542441.2 | XP_011540743.1 | 6 | 913 | 57 | ||
| XM_011542443.2 | XP_011540745.1 | 5 | 931 | 57 | ||
| GenBank mRNA | BC136400.1 | - | 5 | 673 | 9020387 | 57 |
| BC171762.1 | - | 5 | 673 | 57 | ||
| M83554.1 | - | 6 | 854 | 57 | ||
| S75768.1 | - | 6 | 632 | 57 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

TNFRSF8
Genomic Map