| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02033112.hg.1 |
TC02002062.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02033112.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
ENST00000592457.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02033111.hg.1 |
TC02002062.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02033111.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000466417.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000466417.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000466417.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02033113.hg.1 |
TC02002062.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR02033113.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200197477.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200197477.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200197477.hg.1 |
TC0200013331.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000283632.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000466417.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000466417.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000466417.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000471113.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
ENST00000472843.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200008300.hg.1 |
NM_022780.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
ENST00000592457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0200013331.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200197474.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200197474.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200197474.hg.1 |
ENST00000452554.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200197474.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200197474.hg.1 |
TC0200013331.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR0200197476.hg.1 |
ENST00000592457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200197476.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200197476.hg.1 |
ENST00000592457.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0200197476.hg.1 |
NR_026846.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0200197476.hg.1 |
TC0200013331.hg.1 |
Clariom D TC |