Assay Details
| Gene Symbol: TPM3P9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_003148.3 | 2 | 750 | 118 | ||
| GenBank mRNA | AK094777.1 | - | 4 | 690 | 118 | |
| BC061910.1 | - | 1 | 561 | 118 | ||
| BX647962.1 | - | 2 | 737 | 118 | ||
More Information
Set Membership: |
|

TPM3P9
Genomic Map