Assay Details
| Gene Symbol: MT-ND3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | NC_012920.ND3.0 | - | 118 | 150 | ||
| Gene Symbol: SH3KBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AY423734.1 | - | 3353 | 150 | ||
More Information
Additional Information:
NC_012920 (strand:+; 10059..10404); mitochondrially encoded NADH dehydrogenase 3
Set Membership: |
|

MT-ND3
Genomic Map