Assay Details
| Gene Symbol: SSBP3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_103541.1 | 1 | 2559 | 90 | ||
| GenBank mRNA | AF176918.1 | - | 1 | 966 | 90 | |
More Information
Updated Mapping Information
Set Membership: |
|

SSBP3-AS1
Genomic Map