Assay Details
| Gene Symbol: ANKRD20A5P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC022023.1 | - | 3 | 1458 | 4824658 | 137 |
More Information
Updated Mapping Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ANKRD20A5P| Gene Symbol: ANKRD20A5P | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC022023.1 | - | 3 | 1458 | 4824658 | 137 |
Set Membership: |
|