| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000424811.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000522781.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
ENST00000614311.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000012213.hg.1 |
NM_001039762.2 |
Refseq |