Assay Details
Gene Symbol: LCA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_130768.1 | 6 - 7 | 899 | 57 | ||
GenBank mRNA | AF514420.1 | - | 6 - 7 | 901 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3298473 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 15671 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | 8170788 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8170791 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8170789 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8170787 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8170792 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8170790 | - | - |
Affymetrix Human Gene 1.0 ST Array | 8170793 | - | - |
Affymetrix Human Gene 1.1 ST Array | 8170786 | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0X00008804.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00160096.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00160096.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00160096.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00160096.hg.1 | ENST00000614970.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00160096.hg.1 | ENST00000618311.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0X00160096.hg.1 | NR_130768.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0X00160096.hg.1 | TC0X00008804.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1792031 | - | - |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1792031 | - | - |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2067453 | AF514420.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1792031 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2067453 | AF514420.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1792031 | - | - |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2067453 | AF514420.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1792031 | - | - |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2067453 | AF514420.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1792031 | - | - |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2067453 | AF514420.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1792031 | - | - |
