Assay Details
Gene Symbol: LINC00273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_038368.1 | 1 | 1402 | 74 | ||
GenBank mRNA | AY587847.1 | - | 1 | 1402 | 74 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC16001076.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988114 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988117 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16890319 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988124 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988113 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988115 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526421 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988123 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16890298 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526399 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988120 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526419 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988119 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16890304 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P30526468 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P09988121 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16890256 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P16890355 | - | - |
Agilent SurePrint G3 Human CGH Microarray 4x180K | 113838 | - | - |
Agilent SurePrint G3 Human CGH Microarray 8x60K | 23741 | - | - |
Agilent SurePrint G3 Human High-Resolution Discovery Microarray 1x1M | 726309 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 261066 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 16606 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3405916 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 112518 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P03136396 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P03136396 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC1600007647.hg.1 | ENST00000385251.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC2200006438.hg.1 | ENST00000613866.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16015149.hg.1 | ENST00000539813.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR16015149.hg.1 | TC16001076.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | TC1600010122.hg.1 | ENST00000539813.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TCUn_KI270442v100006436.hg.1 | ENST00000617274.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600010122.hg.1 | ENST00000539813.1 | Ensembl |
Affymetrix Human Clariom D Assay | TCUn_KI270442v100006436.hg.1 | ENST00000617274.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC1600007647.hg.1 | ENST00000385251.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC2200006438.hg.1 | ENST00000613866.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600178141.hg.1 | ENST00000539813.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600178141.hg.1 | ENST00000539813.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR1600178141.hg.1 | TC1600010122.hg.1 | Clariom D TC |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1652170 | AY587847.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1652170 | NR_038368.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2211910 | AY587847.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_2211910 | NR_038368.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1652170 | AY587847.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1652170 | NR_038368.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1652170 | AY587847.1 | human_transcriptome |
Illumina human-6 v2.0 expression beadchip | ILMN_1652170 | NR_038368.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2211910 | AY587847.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_2211910 | NR_038368.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1652170 | AY587847.1 | human_transcriptome |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1652170 | NR_038368.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2211910 | AY587847.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_2211910 | NR_038368.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1652170 | AY587847.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1652170 | NR_038368.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2211910 | AY587847.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_2211910 | NR_038368.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1652170 | AY587847.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1652170 | NR_038368.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2211910 | AY587847.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_2211910 | NR_038368.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1652170 | AY587847.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1652170 | NR_038368.1 | human_transcriptome |
