Assay Details
| Gene Symbol: KIFAP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204514.1 | NP_001191443.1 | 17 - 18 | 2330 | 97 | |
| NM_001204516.1 | NP_001191445.1 | 15 - 16 | 2034 | 97 | ||
| NM_001204517.1 | NP_001191446.1 | 16 - 17 | 3275 | 97 | ||
| NM_014970.3 | NP_055785.2 | 16 - 17 | 2166 | 97 | ||
| XM_005244970.1 | XP_005245027.1 | 16 - 17 | 2169 | 97 | ||
| XM_011509307.2 | XP_011507609.2 | 16 - 17 | 2169 | 97 | ||
| GenBank mRNA | AK026471.1 | - | 16 - 17 | 1945 | 97 | |
| AK122897.1 | - | 16 - 17 | 3277 | 97 | ||
| AK297368.1 | - | 15 - 16 | 1961 | 97 | ||
| AK301912.1 | - | 9 - 10 | 1133 | 97 | ||
| AK303052.1 | - | 17 - 18 | 2287 | 97 | ||
| AK315586.1 | - | 16 - 17 | 2080 | 97 | ||
| BC028679.2 | - | 16 - 17 | 2123 | 4829354 | 97 | |
| BC048012.1 | - | 15 - 16 | 2078 | 97 | ||
| U59919.1 | - | 16 - 17 | 2083 | 97 | ||
More Information
Set Membership: |
|

KIFAP3
Genomic Map