Assay Details
| Gene Symbol: SHFM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006304.1 | NP_006295.1 | 1 - 2 | 205 | 77 | |
| GenBank mRNA | AK291070.1 | - | 1 - 2 | 185 | 77 | |
| AK309241.1 | - | 1 - 2 | 163 | 77 | ||
| BC032782.1 | - | 1 - 2 | 165 | 5166837 | 77 | |
| CR456887.1 | - | 1 - 2 | 77 | 77 | ||
| U41515.1 | - | 1 - 2 | 205 | 77 | ||
More Information
Set Membership: |
|

SHFM1
Genomic Map