Assay Details
| Gene Symbol: RNF11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_014372.4 | NP_055187.1 | 1 - 2 | 605 | 71 | |
| GenBank mRNA | AB024703.1 | - | 1 - 2 | 248 | 71 | |
| AF151881.1 | - | 1 - 2 | 248 | 71 | ||
| AK293047.1 | - | 3 - 4 | 434 | 71 | ||
| AK313140.1 | - | 1 - 2 | 367 | 71 | ||
| BC020964.2 | - | 1 - 2 | 200 | 3917224 | 71 | |
| BC047654.1 | - | 1 - 2 | 573 | 5261966 | 71 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000624.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
ABI
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01010317.hg.1 | TC01000624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010317.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010317.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000486691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000486691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000486691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000486691.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010319.hg.1 | TC01000624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010319.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010319.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010319.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010322.hg.1 | TC01000624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010322.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010322.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010322.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2000008448.hg.1 | ENST00000427702.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010321.hg.1 | TC01000624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010321.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010321.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010321.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800006443.hg.1 | ENST00000522979.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015220.hg.1 | ENST00000392337.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015220.hg.1 | ENST00000392337.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010318.hg.1 | TC01000624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010318.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010318.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010324.hg.1 | TC01000624.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010324.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01010324.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158994.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158994.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158994.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158994.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158994.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158994.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158994.hg.1 | TC0100008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100158995.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158995.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158995.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158995.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158995.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158995.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158995.hg.1 | TC0100008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC2000008448.hg.1 | ENST00000427702.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800006443.hg.1 | ENST00000522979.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158991.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158991.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158991.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158991.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158991.hg.1 | TC0100008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100158993.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158993.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158993.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158993.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158993.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158993.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158993.hg.1 | TC0100008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0100158997.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158997.hg.1 | ENST00000486691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158997.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158997.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158997.hg.1 | ENST00000486691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158997.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158997.hg.1 | TC0100008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200015220.hg.1 | ENST00000392337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015220.hg.1 | ENST00000392337.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158992.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158992.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158992.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100158992.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100158992.hg.1 | TC0100008271.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000242719.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000486691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000486691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000486691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000486691.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | ENST00000494873.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100008271.hg.1 | NM_014372.4 | Refseq |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Human Genome Survey Microarray Version 2.0 | 198718 | - | - |

RNF11
Genomic Map