Assay Details
| Gene Symbol: IDS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000202.7 | NP_000193.1 | 2 - 3 | 462 | 72 | |
| NM_006123.4 | NP_006114.1 | 2 - 3 | 462 | 72 | ||
| NR_104128.1 | 2 - 3 | 462 | 72 | |||
| GenBank mRNA | AF050145.1 | - | 2 - 3 | 263 | 72 | |
| BC006170.1 | - | 2 - 3 | 320 | 4025980 | 72 | |
| DA325538.1 | - | 2 - 3 | 462 | 72 | ||
| DB500211.1 | - | 2 - 3 | 447 | 72 | ||
| L40586.1 | - | 2 - 3 | 372 | 72 | ||
| M58342.1 | - | 2 - 3 | 369 | 72 | ||
More Information
Set Membership: |
|

IDS
Genomic Map