Assay Details
| Gene Symbol: ST6GALNAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001160011.1 | NP_001153483.1 | 1 - 2 | 177 | 87 | |
| NM_152996.2 | NP_694541.2 | 1 - 2 | 177 | 87 | ||
| GenBank mRNA | AK091215.1 | - | 1 - 2 | 162 | 87 | |
| AY358540.1 | - | 1 - 2 | 136 | 87 | ||
| BC059363.1 | - | 1 - 2 | 148 | 30341233 | 87 | |
| DB473741.1 | - | 1 - 2 | 178 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK091215.1, AY358540.1, BC059363.1, NM_001160011.1, NM_152996.2 transcripts.
Set Membership: |
|

ST6GALNAC3
Genomic Map