Assay Details
| Gene Symbol: SYT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175733.3 | NP_783860.1 | 3 - 4 | 1283 | 76 | |
| XM_011519900.2 | XP_011518202.1 | 3 - 4 | 1261 | 76 | ||
| XM_011519901.2 | XP_011518203.1 | 3 - 4 | 1261 | 76 | ||
| XM_011519902.2 | XP_011518204.1 | 3 - 4 | 1034 | 76 | ||
| XM_011519904.2 | XP_011518206.1 | 3 - 4 | 1261 | 76 | ||
| GenBank mRNA | AK315013.1 | - | 3 - 4 | 1226 | 76 | |
| BC046367.1 | - | 3 - 4 | 1283 | 5770087 | 76 | |
More Information
Set Membership: |
|

SYT9
Genomic Map