Assay Details
| Gene Symbol: CLDN10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_182848.3 | NP_878268.1 | 1 - 2 | 439 | 74 | |
| GenBank mRNA | AK055855.1 | - | 1 - 2 | 434 | 74 | |
| DA636757.1 | - | 1 - 2 | 440 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC13000335.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR13003469.hg.1 | ENST00000376873.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003469.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003469.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003469.hg.1 | TC13000335.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003470.hg.1 | ENST00000376873.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003470.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR13003470.hg.1 | TC13000335.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152229.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152229.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152229.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152229.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152229.hg.1 | TC1300007752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300152228.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152228.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152228.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152228.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152228.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152228.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152228.hg.1 | TC1300007752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1300152227.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152227.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152227.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152227.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300152227.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152227.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300152227.hg.1 | TC1300007752.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000299339.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376855.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | ENST00000376873.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_001160100.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_006984.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300007752.hg.1 | NM_182848.3 | Refseq |

CLDN10
Genomic Map