Assay Details
| Gene Symbol: RORC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001001523.1 | NP_001001523.1 | 1 - 2 | 148 | 93 | |
| GenBank mRNA | AK097994.1 | - | 1 - 2 | 162 | 93 | |
| AL834219.1 | - | 1 - 2 | 148 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK097994.1, AL834219.1, NM_001001523.1 transcripts.
Set Membership: |
|

RORC
Genomic Map